Skip to Content
MilliporeSigma

BDPH9BCDDFDA

Bide Pharmatech

Methyl 1-chloroisoquinoline-4-carboxylate

Synonym(s):

methyl 1-chloro-4-isoquinolinecarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H8O2N1Cl1
CAS Number:
Molecular Weight:
221.64
MDL number:


Inchi Code

1S/C11H8ClNO2/c1-15-11(14)9-6-13-10(12)8-5-3-2-4-7(8)9/h2-6H,1H3

Inchi Key

YTJDBMFLRYQTPT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library