Skip to Content
MilliporeSigma

BDPH9BCDF2FA

Bide Pharmatech

1-(2-Chlorophenyl)butane-1,3-dione

Synonym(s):

1-(2-chlorophenyl)-1,3-butanedione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9O2Cl1
CAS Number:
Molecular Weight:
196.63
MDL number:


Inchi Code

1S/C10H9ClO2/c1-7(12)6-10(13)8-4-2-3-5-9(8)11/h2-5H,6H2,1H3

Inchi Key

CYPYOGOPKDBEFY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Pale yellow to yellow liquid



Still not finding the right product?

Explore all of our products under 1-(2-Chlorophenyl)butane-1,3-dione




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library