Inchi Code
1S/C9H14F2O4/c1-3-14-7(12)5-9(10,11)6-8(13)15-4-2/h3-6H2,1-2H3
Inchi Key
OJVISWAFWNHSSI-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Liquid
Looking for similar products? Visit Product Comparison Guide
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
