Skip to Content
MilliporeSigma

BDPH9C021B3C

Bide Pharmatech

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine palladium(II)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View



Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Form: solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library