Inchi Code
1S/C9H11NO3/c1-5-7(4-11)8(9(12)13)6(2)10(5)3/h4H,1-3H3,(H,12,13)
Inchi Key
FXVSUGBIKIXSGV-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Powder or crystals
Still not finding the right product?
Explore all of our products under 4-Formyl-1,2,5-trimethyl-1H-pyrrole-3-carboxylic acid
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
