Inchi Code
1S/C29H31N3S/c1-21(25-20-12-18-22-13-10-11-19-26(22)25)30-29(33)31-27(23-14-6-4-7-15-23)28(32(2)3)24-16-8-5-9-17-24/h4-21,27-28H,1-3H3,(H2,30,31,33)/t21-,27+,28+/m0/s1
Inchi Key
FXSOKGPCLADBQO-GJJMEYSYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 1-((1R,2R)-2-(Dimethylamino)-1,2-diphenylethyl)-3-((S)-1-(naphthalen-1-yl)ethyl)thiourea
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
