Inchi Code
1S/C15H20O2/c1-2-11-3-5-12(6-4-11)13-7-9-14(10-8-13)15(16)17/h7-12H,2-6H2,1H3,(H,16,17)/t11-,12-
Inchi Key
DUJHWZDXZFNEKU-HAQNSBGRSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Form: Crystal - Powder / Colour: White - Almost white
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
