Inchi Code
1S/C10H12O4/c1-7(10(12)13-2)14-9-5-3-8(11)4-6-9/h3-7,11H,1-2H3/t7-/m1/s1
Inchi Key
UUYSCNGPNOYZMC-SSDOTTSWSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Form: Crystal - Powder / Colour: White - Pale yellow
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
