Inchi Code
1S/C16H15NO2/c18-14-11-17-15(12-7-3-1-4-8-12)16(19-14)13-9-5-2-6-10-13/h1-10,15-17H,11H2/t15-,16+/m1/s1
Inchi Key
LTPOSIZJPSDSIL-CVEARBPZSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under (5R,6S)-5,6-Diphenyl-2-morpholinone
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
