Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

BDPH9C021FFD

Bide Pharmatech

(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(6-oxo-1H-purin-9(6H)-yl)tetrahydrofuran-3,4-diyl diacetate

Synonym(s):

(2R,3R,4R,5R)-2-(acetoxymethyl)-5-(6-hydroxy-9H-purin-9-yl)tetrahydrofuran-3,4-diyl diacetate

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C16H18O8N4
CAS Number:
Molecular Weight:
394.34
MDL number:

Inchi Code

1S/C16H18N4O8/c1-7(21)25-4-10-12(26-8(2)22)13(27-9(3)23)16(28-10)20-6-19-11-14(20)17-5-18-15(11)24/h5-6,10,12-13,16H,4H2,1-3H3,(H,17,18,24)/t10-,12-,13-,16-/m1/s1

Inchi Key

SFEQTFDQPJQUJM-XNIJJKJLSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to off-white solid

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library