Inchi Code
1S/C27H22N2O/c1-4-11-20(12-5-1)25-19-30-27(29-25)24-18-10-17-23(28-24)26(21-13-6-2-7-14-21)22-15-8-3-9-16-22/h1-18,25-26H,19H2/t25-/m1/s1
Inchi Key
PUSBILBZRCBYNW-RUZDIDTESA-N
Shipping Temperature
Normal
Purity
97%99%
Storage temp.
Keep in dark place, inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under (S)-2-(6-Benzhydrylpyridin-2-yl)-4-phenyl-4,5-dihydrooxazole
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
