Product Name
PCB No 153 solution 10 μg/mL in isooctane, analytical standard
Product Number
36904
Product Brand
sial
CAS Number
35065-27-1
Molecular Weight
360.88
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
CONCENTRATION COMP1~ 10 ug/ml
SOLVENTISOOCTANE
PURITY (GC AREA %)≥ 95.0 %
IDENTIFICATION (GC-MS)IDENTITY CORRESPONDS
REFRACTIVE INDEX N20/D1.3894 - 1.3934
EXPIRY PERIOD60 MONTHS