Product Name
Dextran sulfate sodium salt from Leuconostoc spp. Mr ~500,000
Product Number
31395
Product Brand
sigma
CAS Number
9011-18-1
Molecular Weight
Test
Specification
APPEARANCE (COLOR)White to Faint Beige
APPEARANCE (FORM)Powder or Crystals
SULFUR CONTENT (S/C-ANALYSIS)dry basis
INFRARED SPECTRUMCONFORMS TO STRUCTURE
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
CALCIUM (ICP)≤ 200 mg/kg
CADMIUM (ICP)≤ 5 mg/kg
COBALT (ICP)≤ 5 mg/kg
CHROMIUM (ICP)≤ 5 mg/kg
COPPER (ICP)≤ 5 mg/kg
IRON (ICP)≤ 5 mg/kg
POTASSIUM (ICP)≤ 1000 mg/kg
MAGNESIUM (ICP)≤ 100 mg/kg
MANGANESE (ICP)≤ 5 mg/kg
NICKEL (ICP)≤ 5 mg/kg
LEAD (ICP)≤ 5 mg/kg
ZINC (ICP)≤ 5 mg/kg
CHLORIDE (CL)≤ 100 mg/kg