Product Name
γ-Aminobutyric acid ≥99%
Product Number
a2129
Product Brand
sigma
CAS Number
56-12-2
Molecular Weight
103.12
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
TITRATION (NT) HCLO4 0.1M≥ 99 %
PURITY (TLC AREA %)≥ 99 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)200MG PLUS 4.0 ML OF WATER
INFRARED SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD84 MONTHS