Product Name
TrueGel3D Hydrogel Kits SLO-DEXTRAN, allow cell recovery, PEG non cell-degradable crosslinker
Product Number
true6
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
TRU-SGD:SH-reactive groups30-36 mmol/L
TRU-PEG:Thiol groups20-24 mmol/L
TRU-B72:pHpH 7,1-7,3 at 20-25 °C