MilliporeSigma
Meloxicam: a reappraisal of pharmacokinetics, efficacy and safety.
Expert opinion on pharmacotherapy (2005-10-04)
Brian J Gates, Trang T Nguyen, Stephen M Setter, Neal M Davies
ABSTRACT
The discovery of two distinct isoenzymes of COX has led to the development and clinical introduction of COX-2 inhibitors with increased selectivity onto the market. Meloxicam is a non-steroidal anti-inflammatory drug (NSAID) of the oxicam class, and is a preferential inhibitor of COX-2, demonstrating effectiveness with anti-inflammatory, analgesic and antipyretic activity. Meloxicam is therapeutically utilised in the management of osteoarthritis and rheumatoid arthritis. Trials have examined the risk of gastrointestinal ulceration of meloxicam when compared with traditional non-specific COX-inhibiting NSAIDs with mixed results; meloxicam seems to have a greater gastrointestinal risk than the highly specific COX-2 NSAIDs. Meloxicam has a plasma half-life of approximately 20 h and is convenient for once daily administration. Neither moderate renal nor hepatic insufficiency significantly alters the pharmacokinetics of meloxicam in short-term studies. Furthermore, dose adjustment is not required in the elderly. Recent drug-drug interaction studies have demonstrated that meloxicam interacts with some medications, including cholestyramine, lithium and some inhibitors of cytochrome P450 -2C9 and -3A4. Consequently, increased clinical vigilance should be maintained when coprescribing some medications with meloxicam. Concentration-dependent therapeutic and toxicological effects have yet to be extensively elucidated for meloxicam. Long-term safety in various organ systems, especially in the heart and vascular system and with concomitant drug administration, remains to be proven. The pharmacokinetics of meloxicam enables once daily application, which increases compliance compared with some shorter acting NSAIDs; however, long-term clinical data clearly demonstrating safety and efficacy advantages are lacking.
Product Description
Meloxicam, European Pharmacopoeia (EP) Reference Standard
USP
Meloxicam, United States Pharmacopeia (USP) Reference Standard