Merck
CLS9903
Corning® CellSTACK®细胞培养室
10 chambers, TC-treated, universal cap, sterile, 4/cs
别名:
Corning container
登录 查看组织和合同定价。
选择尺寸
4 EA
$1,500.00
$1,500.00
请联系客服了解存货情况
关于此项目
NACRES:
NB.13
UNSPSC Code:
41122104
跳至
技术服务
需要帮助?我们经验丰富的科学家团队随时乐意为您服务。
让我们为您提供帮助
产品名称
Corning® CellSTACK®细胞培养室, 10 chambers, TC-treated, universal cap, sterile, 4/cs
material
cap (universal)
sterility
sterile
packaging
pack of 1 ea
case of 4 ea
manufacturer/tradename
Corning 9903
suitability
suitable for (scale-up cell culture)
正在寻找类似产品? 访问 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
CLS3272CLS3319CLS3269
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
manufacturer/tradename
Corning 9903
manufacturer/tradename
Corning 3272
manufacturer/tradename
Corning 3319
manufacturer/tradename
Corning 3269
material
cap (universal)
material
clear polystyrene , high-density polyethylene cap
material
orange high-density polyethylene cap, rectangular bottom flask, short neck
material
clear polystyrene , high-density polyethylene cap, rectangular bottom flask, short neck
packaging
pack of 1 ea
packaging
pack of 1, case of 2
packaging
pack of 1, case of 2
packaging
pack of 1, case of 5
suitability
suitable for (scale-up cell culture)
suitability
suitable for (scale-up cell culture)
suitability
suitable for (scale-up cell culture)
suitability
suitable for (scale-up cell culture)
General description
  • 细胞培养扩增的理想选择
  • 培养室产生的细胞产量在107-109范围内
  • 多个滚瓶或旋转瓶的有用替代品。
  • 高机械强度和结构完整性
  • 在发货前已100%进行泄漏测试
  • 通过γ辐射进行无热原认证和灭菌,依据现代制造方法属于低微粒污染
  • 易于使用的广口设计,带螺纹盖和通气盖
  • 26 mm宽大口径灌装口,可直接进入腔室底部,提供更多无菌转移培养基和培养物选项
Legal Information
CellSTACK is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)