生物源
synthetic
化驗
>99% (TLC)
形狀
powder
包裝
pkg of 1 × 10 mg (810325P-10MG)
製造商/商標名
Avanti Research™ - A Croda Brand
螢光
λex 550 nm; λem 570 nm
脂質類型
fluorescent lipids
運輸包裝
dry ice
儲存溫度
−20°C
SMILES 字串
[Cl-].CN1\C(=C\C=C\C2=[N+](CCCCCC(O)=O)c3ccccc3C2(C)C)C(C)(C)c2ccccc12
一般說明
Cyanine 3, free acid is a near-infrared fluorescent probe that can be used to label biomolecules such as peptides.
法律資訊
Avanti Research is a trademark of Avanti Polar Lipids, LLC