Iupac Name
5-bromo-2-[(4-methoxybenzyl)oxy]pyridine
Inchi Code
1S/C13H12BrNO2/c1-16-12-5-2-10(3-6-12)9-17-13-7-4-11(14)8-15-13/h2-8H,9H2,1H3
InChI key
NCWXAIKSFAEBJX-UHFFFAOYSA-N
storage temp.
Sealed in dry, room temperature
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
Solid
Product Link
https://aobchem.com/5-bromo-2-4-methoxybenzyl-oxy-pyridine-147427.html
Signal Word
Warning
MSDS
https://aobchem.com/5-bromo-2-4-methoxybenzyl-oxy-pyridine-147427.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service