Skip to Content
MilliporeSigma

CIAH999A8FE8

ChemScene LLC (Preferred Partner)

Ethyl 4-chloro-2-iodobenzoate

Synonym(s):

ethyl 4-chloro-2-iodobenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C9H8O2Cl1I1
CAS Number:
Molecular Weight:
310.52
MDL number:

Inchi Code

1S/C9H8ClIO2/c1-2-13-9(12)7-4-3-6(10)5-8(7)11/h3-5H,2H2,1H3

Inchi Key

ATVMURDBWDYSMX-UHFFFAOYSA-N

Purity

98%

Storage temp.

4C, protect from light

Country of Origin

CN

Physical Form

Solid

Flash Point

136.5+-22.3C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library