Skip to Content
MilliporeSigma

FLUH99C96BEF

Fluorochem (Preferred Partner)

Ethyl 1-methyl-1H-indole-2-carboxylate

Sign In to View Organizational & Contract Pricing

Select a Size


Iupac Name

ethyl 1-methyl-1H-indole-2-carboxylate

Inchi Code

1S/C12H13NO2/c1-3-15-12(14)11-8-9-6-4-5-7-10(9)13(11)2/h4-8H,3H2,1-2H3

Inchi Key

FRLLPWADCHCBBY-UHFFFAOYSA-N

storage temp.

Ambient Storage

Purity

98%

Country of Origin

GB

Product Link

https://fluorochem.co.uk/product/F446278

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://sds.fluorochem.co.uk/eng/sds_F446278_en.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library