Iniciar sesión para ver los posibles precios especiales de su organización
Seleccione un Tamaño
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Acerca de este artículo
UNSPSC Code:
41105500
Saltar a
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarleQuality Level
usage
sufficient for 100 purifications
sustainability
Greener Alternative Product
concentration
45-55 mg/mL (Magnetic particle)
test parameters
total test time: 5 min
suitability
suitable for nucleic acid purification
greener alternative category
storage temp.
2-30°C
General description
The Ultra pure MagPrep® Viral isolation kit is designed for highly efficient viral nucleic acid isolation from most biological fluids in both manual or high-throughput automated platforms. The kit contains magnetic silica particles and uniquely formulated buffers optimized for rapid easy handling and maximal yield. Nucleic acid isolation is achieved without the need for Proteinase K or sample heating, and all items are shipped and stored at ambient temperature. All kit components are quality tested to be free of contaminant nucleases and DNA, reducing the likelihood of false positives or negatives in downstream assays. The purified nucleic acid eluate is free of PCR inhibitors such as chaotropic salts or alcohols, allowing for effective amplification in various downstream applications such as RT-qPCR, ddPCR, NGS, and cDNA generation. For research or further manufacturing use only. Not intended for direct use in humans or animals. Non-TSCA use only in US.
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.
Features and Benefits
- Nucleic Acid Isolation
- Manual or high-throughput automated isolations
- RT-qPCR
- Digital PCR
- Biological samples can be used
- Room temperature stable reagents
- DNA free
- DNase free
- RNase free
- NICKase free
- simple and fast protocols
Legal Information
MAGPREP is a registered trademark of Merck KGaA, Darmstadt, Germany
Solo componentes del kit
Referencia del producto
Descripción
- UPMPPART-2ML-KC Ultra pure MagPrep Magnetic Particles 1
- UPMPVRBB-40ML-KC Ultra pure MagPrep Viral RNA Lysis and Binding Buffer 1
- UPMPWB1-60ML-KC Ultra pure MagPrep Wash Buffer 1 1
- UPMPWB2-60ML-KC Ultra pure MagPrep Wash Buffer 2 1
- UPMPEB-10ML-KC Ultra pure MagPrep Elution Buffer 1
related product
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?
Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.
Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.
Póngase en contacto con el Servicio técnico