MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Effects of sodium dodecyl sulphate on enhancement of lipoxygenase activity of hemoglobin.
Indian journal of experimental biology (2013-08-31)
F C Ezebuo, S O O Eze, F C Chilaka
RESUMEN
Lipoxygenases comprise a family of non-heme iron-containing enzymes that catalyze the stereospecific dioxygenation of polyunsaturated fatty acids with 1, 4-cis-cis-pentadiene structure. Hemoglobin, a heme iron-containing protein has been reported to have lipoxygenase activity but the assay conditions that could enhance the activity remain obscure. Therefore, establishment of optimum assay conditions for lipoxygenase activity of hemoglobin could allow modeling of hemoglobin as lipoxygenase. Hemoglobin was extracted from blood of an identified individual of genotype AA. The hemoglobin was dialyzed at 4 degrees C for 24 h against 50 mM Tris-HCl buffers (pH 8.5 and 7.2) and effects of sodium dodecyl sulphate (SDS) and linoleic studied at pH 5.0 and 7.2 with UV-VIS Titration Spectrophotometry. The results show that 3.3, 8.6 and 88.1% concentrations of met-hemoglobin were found in presence of 0.0 mM SDS at pH 5.0 and 7.2, 1.043 mM SDS at pH 7.2 and 0.404 mM SDS at pH 5.0 respectively. Also, the difference spectra of hemoglobin in presence of linoleic acid showed positive peak at 285 nm which suggest the presence of oxodienes--a reaction product of hydroperoxidase activity of lipoxygenase. Formation of met-hemoglobin/met-myoglobin is highly correlated with lipid oxidation. Since highest concentration of met-hemoglobin (88.1%) was observed in presence of 0.404 mM SDS at pH 5.0, lipoxygenase activity of hemoglobin was enhanced in presence of SDS under these conditions.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dodecilsulfatosódico, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Dodecilsulfatosódico, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Dodecilsulfatosódico, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Hemoglobin human, lyophilized powder
Sigma-Aldrich
Dodecilsulfatosódico, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Dodecilsulfatosódico, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Hemoglobin from bovine blood, lyophilized powder
Sigma-Aldrich
Ácido linoleico, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Hemoglobin from bovine blood, suitable for protease substrate, substrate powder
Supelco
Dodecilsulfatosódico, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Dodecilsulfatosódico, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Dodecilsulfatosódico, ≥98.0% (GC)
Sigma-Aldrich
Dodecilsulfatosódico, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Hemoglobin from bovine blood, dried erythrocytes
Sigma-Aldrich
Ácido linoleico, technical, 58-74% (GC)
Supelco
Ácido linoleico, analytical standard
Sigma-Aldrich
Linoleic acid sodium salt, ≥97% (GC)
Sigma-Aldrich
Dodecilsulfatosódico, ≥90% ((Assay))
Sigma-Aldrich
Dodecilsulfatosódico, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Hemoglobin porcine, lyophilized powder
Millipore
Hemoglobin from bovine blood, from bovine blood, powder, for Neisseria spp., pack of 100 g or 500 g, suitable for microbiology
Sigma-Aldrich
Dodecilsulfatosódico, SAJ special grade, ≥97.0%
Sigma-Aldrich
Dodecilsulfatosódico, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Dodecilsulfatosódico, ≥99.0%