MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
NAIF1 inhibits gastric cancer cells migration and invasion via the MAPK pathways.
Journal of cancer research and clinical oncology (2014-11-30)
Mei Yang, Yu-Yu Gu, Hua Peng, Mei Zhao, Jia Wang, Sheng-Kai Huang, Xing-Hua Yuan, Jia Li, Jian-Li Sang, Qing Luo, Changzhi Huang
RESUMEN
Nuclear apoptosis-inducing factor 1 (NAIF1) could induce apoptosis in gastric cancer cells. Previously, we have reported that the expression of NAIF1 protein is down-regulated in gastric cancer tissues compared with the adjacent normal tissues. However, the role of NAIF1 in gastric cancer cells is not fully understood. The effects of NAIF1 on cell viability were evaluated by MTT and colony formation assays. The ability of cellular migration and invasion were analyzed by transwell assays. The expression levels of targeted proteins were determined by western blot. The relative RNA expression levels were analyzed using quantitative polymerase chain reaction assays. Xenograft experiment was employed to determine the anti-tumor ability of NAIF1 in vivo. The study demonstrates that transient transfection of NAIF1 in gastric cancer cells BGC823 and MKN45 could inhibit the cell proliferation, migration, and invasion of the two gastric cancer cell lines. The tumor size is smaller in NAIF1-overexpressed MKN45 cell xenograft mice than in unexpressed group. Further in-depth analysis reveals that NAIF1 reduces the expression of MMP2 as well as MMP9, and inhibits the activation of FAK, all of which are key molecules involved in regulating cell migration and invasion. In addition, NAIF1 inhibits the expression of c-Jun N-terminal kinase (JNK) by accelerating its degradation through ubiquitin-proteasome pathway. Meanwhile, NAIF1 reduces the mRNA and protein expression of ERK1/2. Our study revealed that NAIF1 plays a role in regulating cellular migration and invasion through the MAPK pathways. It could be a therapeutic target for gastric cancer.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dodecilsulfatosódico, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Roche
Reactivo de transfección de ADN X-tremeGENE HP, High-performance polymer reagent for transfecting many cell lines
Sigma-Aldrich
Dodecilsulfatosódico, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Azul de tiazolil Bromuro de tetrazolio, 98%
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Dodecilsulfatosódico, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Fluoruro de fenilmetansulfonilo, ≥98.5% (GC)
Sigma-Aldrich
Azul de tiazolil Bromuro de tetrazolio, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dodecilsulfatosódico, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Dodecilsulfatosódico, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Fluoruro de fenilmetansulfonilo, ≥99.0% (T)
Supelco
Dodecilsulfatosódico, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Cloruro de sodio, JIS special grade, ≥99.5%
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Sigma-Aldrich
Dodecilsulfatosódico, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Dodecilsulfatosódico, ≥98.0% (GC)
Sigma-Aldrich
Dodecilsulfatosódico, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Cloruro de sodio, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%