MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Multidimensional nanofibrous scaffolds of poly(lactide-co-caprolactone) and poly(ethyl oxazoline) with improved features for cardiac tissue engineering.
Multidimensional nanofibrous scaffolds of poly(lactide-co-caprolactone) and poly(ethyl oxazoline) with improved features for cardiac tissue engineering.
Nanomedicine (London, England) (2015-11-27)
Rajesh Lakshmanan, Uma Maheswari Krishnan, Swaminathan Sethuraman
RESUMEN
The aim of the study is to develop scaffolds that mimic native tissue properties for effective regeneration of the myocardium, which is affected by the gradual thinning of left ventricular tissue after an infarction. Heterogenous nanofibrous scaffolds made of poly(lactide-co-caprolactone) and poly(ethyl oxazoline) were characterized for physico-chemical properties. The biocompatibility of the scaffolds was evaluated by studying the adhesion, proliferation and differentiation of H9c2 cells. The scaffolds mimic the cardiac extracellular matrix and showed enhanced tensile strength, improved cell compatibility along with the expression of cardiac marker proteins. Our experimental data confirmed the importance of native tissue architecture and mechanical strength for improved cell response in cardiac tissue engineering.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Alcohol etílico puro 200, Molecular Biology
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Alcohol etílico puro, 200 proof
Sigma-Aldrich
Base Trizma®, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Dodecilsulfatosódico, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Base Trizma®, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Acida sódica, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dodecilsulfatosódico, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Dodecilsulfatosódico, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dodecilsulfatosódico, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Tris(hidroximetil)-aminometano, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Base Trizma®, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Acida sódica, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Dodecilsulfatosódico, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
SAFC
Sodium chloride solution, 5 M
Supelco
Dodecilsulfatosódico, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)