EMAIL THIS PAGE TO A FRIEND
Journal of the neurological sciences
Neuroleptic malignant syndrome associated with diphenhydramine and diprophyllin overdose in a depressed patient.
PMID 10064181
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0633
7-(2,3-Dihydroxypropyl)theophylline
C10H14N4O4