EMAIL THIS PAGE TO A FRIEND
Advances in protein chemistry
Pyrroloquinoline quinone (PQQ) from methanol dehydrogenase and tryptophan tryptophylquinone (TTQ) from methylamine dehydrogenase.
PMID 11665494
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7783
Pyrroloquinoline quinone, ≥95.0% (HPLC)
C14H6N2O8
64682
Pyrroloquinoline quinone, ≥97.0% (HPLC)
C14H6N2O8