EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Dipolar cycloaddition of ethyl isocyanoacetate to 3-chloro-2-(methylthio)/2-(methylsulfonyl)quinoxalines: highly regio- and chemoselective synthesis of substituted imidazo[1,5-a]quinoxaline-3-carboxylates.
PMID 17539688
Abstract
An efficient route for regio- and chemoselective synthesis of substituted 3-(carboethoxy)imidazo[1,5-a]quinoxalines and novel diimidazo[1,5-a:5',1'-c]quinoxalines via base-induced cycloaddition of ethyl isocyanoacetate to unsymmetrically substituted 3-chloro-2-(methylthio)/2-(methylsulfonyl)quinoxalines has been reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E33300
Ethyl isocyanate, 98%
C3H5NO