EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Cloning and expression of the sucrose phosphorylase gene from Leuconostoc mesenteroides in Escherichia coli.
PMID 18038113
Abstract
The gene encoding sucrose phosphorylase (742sp) in Leuconostoc mesenteroides NRRL B-742 was cloned and expressed in Escherichia coli. The nucleotide sequence of the transformed 742sp comprised an ORF of 1,458 bp giving a protein with calculated molecular mass of 55.3 kDa. 742SPase contains a C-terminal amino acid sequence that is significantly different from those of other Leu. mesenteroides SPases. The purified 742SPase had a specific activity of 1.8 U/mg with a K (m) of 3 mM with sucrose as a substrate; optimum activity was at 37 degrees C and pH 6.7. The purified 742SPase transferred the glucosyl moiety of sucrose to cytosine monophosphate (CMP).