EMAIL THIS PAGE TO A FRIEND
Journal of animal science
Effects of natural (RRR alpha-tocopherol acetate) or synthetic (all-rac alpha-tocopherol acetate) vitamin E supplementation on reproductive efficiency in beef cows.
PMID 20495121
Abstract
The objective was to determine the effects of natural- or synthetic-source vitamin E on reproductive efficiency in Angus-cross beef cows. In Exp. 1, one hundred fifty-two cows were fed hay and corn silage based diet and assigned to 1 of 3 dietary supplements (3 pens/treatment): 1) containing no additional vitamin E (CON), 2) formulated to provide 1,000 IU x d(-1) of synthetic-source vitamin E (SYN; all-rac or dl-alpha-tocopherol acetate), or 3) formulated to provide 1,000 IU x d(-1) of natural-source vitamin E (NAT; RRR or D-alpha-tocopherol acetate). In Exp. 2, seventy-five cows (2 reps/treatment) were assigned to similar treatments as Exp. 1; however, a vitamin-mineral supplement was offered for ad libitum intake and vitamin intake was calculated from predicted mineral intakes. Cows grazed pastures rather than being fed hay and corn silage as in Exp. 1. In Exp. 1 and 2, supplementation began 6 wk prepartum and continued until initiation of the breeding season. Blood samples were collected at calving (Exp. 1) or breeding (Exp. 2) to determine alpha-tocopherol concentration and weekly beginning 4 wk postpartum (Exp. 1) or 7 and 14 d before estrus synchronization (Exp. 2) to determine return to estrus via progesterone concentration. Cows were synchronized and bred by AI based on heat detection; nonresponding cows were time bred (AI) 66 h after PGF(2 alpha) injection, and cows returning to estrus after AI were bred by natural service. In Exp. 1, cows supplemented with NAT and SYN had greater (P < 0.001) serum concentrations of alpha-tocopherol at calving compared with CON cows. Dietary supplement did not affect (P >or= 0.55) the percentage of cows cycling before synchronization or the number of days to return to estrus by cows that resumed estrus before synchronization. Cows supplemented with SYN tended to have greater first service conception rates compared with CON and NAT (P = 0.09); however, first plus second services combined and overall conception rates were not affected (P >or= 0.23). In Exp. 2, NAT cows had greater (P = 0.002) concentrations of alpha-tocopherol at breeding, whereas there was no difference (P > 0.05) between SYN and CON. Supplementation of SYN or NAT did not affect (P >or= 0.17) days to resumption of estrus before breeding, first service, first plus second services combined, or overall conception rates. These data suggest that supplementation of SYN or NAT source vitamin E increased alpha-tocopherol concentration in cows; however, effects on reproductive efficiency are minimal.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3376
DL-α-Tocopherol acetate, ≥96% (HPLC)
C31H52O3