EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Microwave-assisted synthesis of hydrophilic BaYF5:Tb/Ce,Tb green fluorescent colloid nanocrystals.
PMID 21076744
Abstract
Hydrophilic Ce, Tb doped BaYF(5) nanocrystals with uniform size were synthesized by a microwave-assisted route. The synthesized nanocrystals can be well dispersed in hydrophilic solutions (DMSO, DMF, EG, H(2)O). This synthesis procedure represents a less time consuming method, with high product yield and without using any assistant or/and template reagents, which may be expected to be a general method for rapid synthesis of other hydrophilic RE doped fluoride fluorescent nanocrystals. The Ce(3+), Tb(3+) codoped BaYF(5) nanocrystals show bright green fluorescence emission. The Ce(3+) acts as an effective energy transfer medium and the emission at the high (5)D(3) energy level of Tb is enhanced in this host material.