EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Ultrasound-mediated destabilization and drug release from liposomes comprising dioleoylphosphatidylethanolamine.
PMID 21238586
Abstract
Novel sonosensitive doxorubicin-containing liposomes comprising dioleoylphosphatidylethanolamine (DOPE) as the main lipid constituent were developed and characterized in terms of ultrasound-mediated drug release in vitro. The liposome formulation showed high sonosensitivity; where approximately 95% doxorubicin was released from liposomes after 6min of 40kHz US exposure in buffered sucrose solution. This represented a 30% increase in release extent in absolute terms compared to liposomes comprising the saturated lipid analogue distearoylphosphatidylethanolamine (DSPE), and a 9-fold improvement in release extent when compared to standard pegylated liposomal doxorubicin, respectively. Ultrasound release experiments in the presence of serum showed a significantly reduction in sonosensitivity of DSPE-based liposomes, whilst the release properties of DOPE-based liposomes were essentially maintained. Dynamic light scattering measurements and cryo-transmission electron microscopy of DOPE-based liposomes after ultrasound treatment indicated liposome disruption and formation of various lipid structures, corroborating the high release extent. The results point to the potential of DOPE-based liposomes as a new class of drug carriers for ultrasound-mediated drug delivery.