EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Bioseparation of recombinant proteins from plant extract with hydrophobin fusion technology.
PMID 22160918
Abstract
Two main hurdles hinder the widespread acceptance of plants as a preferred protein expression platform: low accumulation levels and expensive chromatographic purification methods. Fusion of proteins of interest to fungal hydrophobins has provided a tool to address both accumulation and purification issues. In this method, we describe the one-step purification of a GFP-HFBI fusion from crude plant extract using an aqueous two-phase system (ATPS). ATPS can be carried out in a very short time frame, yields relatively pure protein with very few contaminants, and does not require any chromatographic column steps. This purification system takes advantage of the affinity of hydrophobins to the micellar phase of widely available nonionic surfactants, such as Triton X-114, and can be easily scaled up for industrial-scale protein purification.
Related Materials