EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Optical spectroscopic exploration of binding of Cochineal Red A with two homologous serum albumins.
PMID 22397587
Abstract
Cochineal Red A is a negatively charged synthetic azo food colorant and a potential carcinogen. We present here the study of binding of Cochineal Red A with two homologous serum albumins, human (HSA) and bovine (BSA), in aqueous pH 7.4 buffer by optical spectroscopic techniques. Protein intrinsic fluorescence quenching by Cochineal Red A occurs through ground-state static interaction and its binding with BSA is stronger than with HSA. The magnitudes of thermodynamic parameters suggest that dye binding occurs principally via electrostatic complexation. Site-marker competitive binding shows that Cochineal Red A binds primarily to site I of serum albumins. Circular dichroic spectra indicate that dye binding results in some conformational modification of serum albumins. Increased ionic strength of the medium results in lowering of binding. This study provides an important insight into possible means of removal of dye toxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199737
New Coccine, Dye content 75 %
C20H11N2Na3O10S3
18137
Ponceau 4R, analytical standard
C20H11N2Na3O10S3