EMAIL THIS PAGE TO A FRIEND
Organic letters
Total synthesis of the terpenoid buddledone A: 11-membered ring-closing metathesis.
PMID 22432972
Abstract
The first total synthesis of buddledone A was accomplished in seven steps from methyl ethyl ketone (MEK). The key step in the sequence featured an 11-membered ring formation by ring-closing metathesis.