EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Cobalt-catalyzed direct arylation and benzylation by C-H/C-O cleavage with sulfamates, carbamates, and phosphates.
PMID 22791393