EMAIL THIS PAGE TO A FRIEND
Molecular pharmaceutics
Eudragit E100 as a drug carrier: the remarkable affinity of phosphate ester for dimethylamine.
PMID 22808998
Abstract
Therapeutic agents containing phosphate groups in their molecules have increasing therapeutic impact. The object of this study was to characterize the cationic polyelectrolyte Eudragit E100 (EuE100) as a carrier for drugs containing phosphate groups, using dexamethasone phosphate (DP) as a model. A series of EuE100-DP complexes was obtained by acid-base reaction in which DP neutralized 12.5-75% of the basic groups of EuE100. The solids obtained after solvent evaporation revealed by spectroscopic characterization the complete reaction between the components through the ionic interaction between the amine groups of EuE100 and the phosphate groups of DP. The reversibility of the counterion condensation, evaluated through the proton-withdrawing effect produced by the ionic exchange generated by titration with NaCl, showed a remarkable high affinity between EuE100 and DP. In line, drug delivery in bicompartimental Franz cells toward water as receptor medium was very slow (2% in 6 h). However, it was increased as water was replaced by NaCl solution, which upon diffusion generates ionic exchange. A sustained release of DP with noticeable zero order kinetics accounted for a remarkable high affinity, mainly due to the electrostatic attraction. The release rate remains constant regardless of the saline concentration of the media. Besides, the delivery control is maintained even in gastric simulated fluid, a property not informed previously for EuE100 complexes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
126365
Dimethylamine hydrochloride, 99%
C2H7N · HCl
38960
Dimethylamine hydrochloride, purum, ≥98.0% (AT)
C2H7N · HCl