EMAIL THIS PAGE TO A FRIEND
Journal of materials science. Materials in medicine
"In vitro" behaviour of adult mesenchymal stem cells of human bone marrow origin seeded on a novel bioactive ceramics in the Ca2SiO4-Ca 3(PO4)2 system.
PMID 22903601
Abstract
This work describes the evaluation of three ceramic materials as potential osteogenic substrate for bone tissue engineering. The capacity of adult human mesenchymal stem cells cultured under experimental conditions known to adhere, proliferate and differentiate into osteoblasts was studied. Two types of culture medium: growth medium and osteogenic medium were evaluated. The materials were pure α-tricalcium phosphate and also αTCP doped with either 1.5 or 3 wt% of dicalcium silicate. The results showed that the hMSCs cultured adhered, spread, proliferated and produced mineralized extracellular matrix on all the ceramics studied. They showed an osteoblastic phenotype, especially in the αTCP doped with 1.5 wt% C(2)S, indicating osteoblastic differentiation as a result of the increased concentration of silicon in solid solution in TCP. Ceramics evaluated in this work are bioactive, cytocompatible and capable of promoting the differentiation of hMSCs into osteoblast.