EMAIL THIS PAGE TO A FRIEND
Journal of environmental sciences (China)
La-EDTA coated Fe3O4 nanomaterial: preparation and application in removal of phosphate from water.
PMID 23596964
Abstract
La-EDTA-Fe3O4 was prepared by a chemical co-precipitation method. The magnetic composite was characterized by transmission electron microscopy (TEM), X-ray diffraction (XRD) and Fourier transform infrared spectroscopy (FT-IR). Furthermore, the adsorption properties of La-EDTA-Fe3O4 toward phosphate in water were investigated. The uptake rate of phosphate in water by La-EDTA-Fe3O4 was 3-1000 times than that of EDTA-Fe3O4, and reached 97.8% at 7 hr. The adsorption process agreed well with the Freundlich model and kinetics studies showed that the adsorption of phosphate proceeds according to pseudo second-order adsorption kinetics. The maximum removal rate was achieved at pH 6.0-7.0. The La-EDTA-Fe3O4 had good adsorption properties and could be separated well from aqueous solution by a permanent magnet. Therefore, this nanomaterial has potential application for the removal of phosphate from large water bodies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ED2SC
Ethylenediaminetetraacetic acid calcium disodium salt
C10H12CaN2Na2O8
PHR1068
Ethylenediaminetetraacetic acid disodium salt dihydrate
C10H14N2Na2O8 · 2H2O