EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Differential modulation of brain nicotinic acetylcholine receptor function by cytisine, varenicline, and two novel bispidine compounds: emergent properties of a hybrid molecule.
PMID 23959137
Abstract
Partial agonist therapies for the treatment of nicotine addiction and dependence depend on both agonistic and antagonistic effects of the ligands, and side effects associated with other nAChRs greatly limit the efficacy of nicotinic partial agonists. We evaluated the in vitro pharmacological properties of four partial agonists, two current smoking cessation drugs, varenicline and cytisine, and two novel bispidine compounds, BPC and BMSP, by using defined nAChR subtypes expressed in Xenopus laevis oocytes and human embryonic kidney 293 cells. Similar to varenicline and cytisine, BPC and BMSP are partial agonists of α4β2 nAChRs, although BMSP produced very little activation of these receptors. Unlike varenicline and cytisine, BPC and BMSP showed desired low activity. BPC produced mecamylamine-sensitive steady-state activation of α4* receptors that was not evident with BMSP. We evaluated the modulation of α4*- and α7-mediated responses in rat lateral geniculate nucleus (LGN) neurons and hippocampal stratum radiatum (SR) interneurons, respectively. The LGN neurons were sensitive to a very low concentration of varenicline, and the SR interneuron responses were also sensitive to varenicline at a submicromolar concentration. Although 300 nM BPC strongly inhibited the ACh-evoked responses of LGN neurons, it did not inhibit the α7 currents of SR interneurons. Similar results were observed with 300 nM BMSP. Additionally, the bispidine compounds were efficacious in the mouse tail suspension test, demonstrating that they affect receptors in the brain when delivered systemically. Our data indicate that BPC and BMSP are promising α4β2* partial agonists for pharmacotherapeutics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
712264
Cytisine, ≥99.0% (HPLC), ≥99%
C11H14N2O
C2899
Cytisine, ≥99%, powder
C11H14N2O