EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
25th anniversary article: Understanding the lithiation of silicon and other alloying anodes for lithium-ion batteries.
PMID 24038172
Abstract
Alloying anodes such as silicon are promising electrode materials for next-generation high energy density lithium-ion batteries because of their ability to reversibly incorporate a high concentration of Li atoms. However, alloying anodes usually exhibit a short cycle life due to the extreme volumetric and structural changes that occur during lithium insertion/extraction; these transformations cause mechanical fracture and exacerbate side reactions. To solve these problems, there has recently been significant attention devoted to creating silicon nanostructures that can accommodate the lithiation-induced strain and thus exhibit high Coulombic efficiency and long cycle life. In parallel, many experiments and simulations have been conducted in an effort to understand the details of volumetric expansion, fracture, mechanical stress evolution, and structural changes in silicon nanostructures. The fundamental materials knowledge gained from these studies has provided guidance for designing optimized Si electrode structures and has also shed light on the factors that control large-volume change solid-state reactions. In this paper, we review various fundamental studies that have been conducted to understand structural and volumetric changes, stress evolution, mechanical properties, and fracture behavior of nanostructured Si anodes for lithium-ion batteries and compare the reaction process of Si to other novel anode materials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
499811
Lithium, granular, 99% trace metals basis
Li
444456
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Li
266000
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Li
265993
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Li
320080
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Li
265985
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Li
265969
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Li
220914
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Li
278327
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Li
GF47159826
Lithium, foil, 25x100mm, thickness 0.6mm, as rolled, 99.9%
Li
GF11639628
Lithium, foil, not light tested, 45x200mm, thickness 0.12mm, as rolled, 99.9%
Li
GF40266682
Lithium, foil, not light tested, 38x500mm, thickness 0.20mm, as rolled, 99.9%
Li
GF62158596
Lithium, foil, not light tested, 38x200mm, thickness 0.20mm, as rolled, 99.9%
Li
343250
Silicon, pieces, 99.95% trace metals basis
Si
267414
Silicon, powder, −60 mesh, 99.999% trace metals basis
Si
215619
Silicon, powder, −325 mesh, 99% trace metals basis
Si
633097
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Si
646687
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Si
647535
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Si
647780
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Si
647802
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Si
647675
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Si
647764
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Si
647101
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Si
647543
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Si
647799
Silicon, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Si
647705
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Si
647772
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Si
767492
Silicon, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.999% trace metals basis
Si
GF37993595
Silicon, rod, 100mm, diameter 10.0mm, single crystal - random orientation, 100%
Si