EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Characterization and preparation of p(U-MMA-An) interpenetrating polymer network damping and absorbing material.
PMID 24738404
Abstract
P(U-MMA-ANI) interpenetrating polymer network (IPN) damping and absorbing material is successfully synthesized by PANI particles served as an absorbing agent with the microemulsion polymerization and P(U-MMA) foam IPN network structure for substrate materials with foaming way. P(U-MMA-ANI) IPN is characterized by the compression mechanical performance testing, TG-DSC, and DSC. The results verify that the P(U-MMA) IPN foam damping material has a good compressive strength and compaction cycle property, and the optimum content of PMMA was 40% (mass) with which the SEM graphs do not present the phase separation on the macro level between PMMA and PU, while the phase separation was observed on the micro level. The DTG curve indicates that because of the formation of P(U-MMA) IPN, the decomposition temperature of PMMA and the carbamate in PU increases, while that of the polyol segment in PU has almost no change. P(U-MMA-ANI) IPN foam damping and absorbing material is obtained by PANI particles served as absorbing agent in the form of filler, and PMMA in the form of micro area in substrate material. When the content of PANI was up to 2.0% (mass), the dissipation factor of composites increased, and with the increasing of frequency the dissipation factor increased in a straight line.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
452130
Poly(methyl methacrylate), isotactic, >80% isotactic