EMAIL THIS PAGE TO A FRIEND
Clinical and experimental medicine
TRB3 mediates renal tubular cell apoptosis associated with proteinuria.
PMID 24925634
Abstract
Proteinuria may contribute to progressive renal damage by inducing tubulointerstitial inflammation, fibrosis and tubular cell apoptosis, but the underlying mechanisms remain largely unknown. TRB3 is a kinase-like molecule that can modify cellular survival and interfere with signal transduction pathways. We seek to determine the role of TRB3 in renal tubular cell apoptosis associated with proteinuria. Herein, we reported that in a rat tubular cell line, high concentration of albumin augmented TRB3 expression and induced apoptosis, while TRB3 silencing with special small interference RNA significantly attenuated apoptosis. In addition, we found that albumin-induced apoptosis was related to inhibition of Akt phosphorylation, which was, however, partially reversed by TRB3 silencing, indicating that TRB3 worked through Akt pathway in this apoptotic signaling cascade. In vivo, we observed increased TRB3 expression in kidneys of streptozotocin-induced diabetic nephropathy model and albumin-overload nephropathy model, both of which showed overt proteinuria. Notably, proteinuria induced apoptosis in renal tubules, which was less severe after genetically inhibition of TRB3. Taken together, these results suggest that TRB3 mediates renal tubular cell apoptosis induced by protein overload, broadening our understanding of the pathogenesis of progressive proteinuric kidney diseases.