EMAIL THIS PAGE TO A FRIEND
Oncology Reports
Apoptosis- and differentiation-inducing activities of jacaric acid, a conjugated linolenic acid isomer, on human eosinophilic leukemia EoL-1 cells.
PMID 25174702