PLoS ONE
Pharmacological activation of the EDA/EDAR signaling pathway restores salivary gland function following radiation-induced damage.
PMID 25409170
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000113
Carbachol, European Pharmacopoeia (EP) Reference Standard
C6H15ClN2O2