EMAIL THIS PAGE TO A FRIEND
Antimicrobial Agents and Chemotherapy
Influenza viral load and peramivir kinetics after single administration and proposal of regimens for peramivir administration against resistant variants.
PMID 25547357