EMAIL THIS PAGE TO A FRIEND
Annals of biomedical engineering
Coronary Flow Impacts Aortic Leaflet Mechanics and Aortic Sinus Hemodynamics.
PMID 25636598
Abstract
Mechanical stresses on aortic valve leaflets are well-known mediators for initiating processes leading to calcific aortic valve disease. Given that non-coronary leaflets calcify first, it may be hypothesized that coronary flow originating from the ostia significantly influences aortic leaflet mechanics and sinus hemodynamics. High resolution time-resolved particle image velocimetry (PIV) measurements were conducted to map the spatiotemporal characteristics of aortic sinus blood flow and leaflet motion with and without physiological coronary flow in a well-controlled in vitro setup. The in vitro setup consists of a porcine aortic valve mounted in a physiological aorta sinus chamber with dynamically controlled coronary resistance to emulate physiological coronary flow. Results were analyzed using qualitative streak plots illustrating the spatiotemporal complexity of blood flow patterns, and quantitative velocity vector and shear stress contour plots to show differences in the mechanical environments between the coronary and non-coronary sinuses. It is shown that the presence of coronary flow pulls the classical sinus vorticity deeper into the sinus and increases flow velocity near the leaflet base. This creates a beneficial increase in shear stress and washout near the leaflet that is not seen in the non-coronary sinus. Further, leaflet opens approximately 10% farther into the sinus with coronary flow case indicating superior valve opening area. The presence of coronary flow significantly improves leaflet mechanics and sinus hemodynamics in a manner that would reduce low wall shear stress conditions while improving washout at the base of the leaflet.