EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Decreased mRNA and protein expression of TWIST1 in myocardial tissue of fetuses with ventricular septal defects.
PMID 25955272
Abstract
Ventricular septal defect (VSD) is the most common type of congenital heart disease (CHD). The single gene mutations or absences that contribute to VSD development are well established; however, the aim of the present study was to measure gene expression variation between VSDs and normal fetal myocardial tissue. TWIST1, an important tumor biomarker, is a basic helix-loop-helix transcription factor that regulates cell proliferation, migration and differentiation in embryonic development and transformed tumor cells. Although growing evidence demonstrates that TWIST1 participates in a variety of human neoplastic diseases, the role of TWIST1 in VSD has remained elusive. Twenty-six VSD fetal myocardial tissue samples and 12 normal samples at matched gestational weeks (22-28 weeks) were included in the present study. Using reverse transcription quantitative polymerase chain reaction (PCR) and real-time PCR, it was demonstrated that TWIST1 mRNA was reduced by almost two-fold in the VSD samples compared with the normal samples. Western blot analysis also revealed that TWIST1 expression was decreased by ~three-fold (P=0.001) in the VSD samples compared with that in the normal samples. Of note, five complete ventricular (also called functionally univentricular or single ventricular) septal ageneses were identified among the specimens. For the five complete ventricular septal agenesis samples, similar results to those for other VSD fetal myocardial tissues were obtained. In conclusion, the results of the present study showed that TWIST1 mRNA and protein levels were reduced in VSDs. The present study was the first, to the best of our knowledge, to report that TWIST1 is not only a tumor biomarker, but may also be involved in the pathogenesis of VSD.