EMAIL THIS PAGE TO A FRIEND
The International Journal of Biochemistry & Cell Biology
Arjunolic acid ameliorates reactive oxygen species via inhibition of p47(phox)-serine phosphorylation and mitochondrial dysfunction.
PMID 26319153