EMAIL THIS PAGE TO A FRIEND
Beilstein journal of organic chemistry
Tetrathiafulvalene-based azine ligands for anion and metal cation coordination.
PMID 26425193
Abstract
The synthesis and full characterization of two tetrathiafulvalene-appended azine ligands, namely 2-([2,2'-bi(1,3-dithiolylidene)]-4-yl)-6-((2,4-dinitrophenyl)hydrazono)methyl)pyridine (L1) and 5-([2,2'-bi(1,3-dithiolylidene)]-4-yl)-2-((2,4-dinitrophenyl)hydrazono)methyl)pyridine (L2) are described. The crystal structure of ligand L1 indicates that the ligand is completely planar with the presence of a strong intramolecular N3-H3···O1 hydrogen bonding. Titration experiments with inorganic anions showed that both ligands are suitable candidates for the sensing of fluoride anions. Ligand L2 was reacted with a Re(I) cation to yield the corresponding rhenium tricarbonyl complex 3. In the crystal structure of the newly prepared electroactive rhenium complex the TTF is neutral and the rhenium cation is hexacoordinated. The electrochemical behavior of the three compounds indicates that they are promising for the construction of crystalline radical cation salts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
525693
6-Bromo-2-pyridinecarboxaldehyde, 97%
C6H4BrNO